Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033105.4 | NP_149096.2 | 4 | 461 | ||
XM_011517618.2 | XP_011515920.1 | 4 | 393 | |||
XM_011517619.2 | XP_011515921.1 | 2 | 205 | |||
XM_011517620.2 | XP_011515922.1 | 4 | 454 | |||
XM_011517621.2 | XP_011515923.1 | 2 | 151 | |||
GenBank mRNA | AF368276.1 | - | 4 | 439 | ||
AL133660.1 | - | 5 | 1947 | |||
BC016742.2 | - | 4 | 458 | 4830917 | ||
BG701336.1 | - | 4 | 478 |