Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003675.3 | NP_003666.1 | 4 | 493 | ||
XM_011519754.2 | XP_011518056.1 | 5 | 618 | |||
XM_017016860.1 | XP_016872349.1 | 4 | 621 | |||
GenBank mRNA | AK314814.1 | - | 4 | 468 | ||
AU142253.1 | - | 4 | 493 | |||
BC000794.1 | - | 4 | 415 | 3451639 | ||
U51990.1 | - | 4 | 417 |