Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003689.3 | NP_003680.2 | 3 | 539 | ||
GenBank mRNA | AF026947.1 | - | 3 | 507 | ||
AK225267.1 | - | 3 | 510 | |||
AK310609.1 | - | 3 | 496 | |||
BC004111.2 | - | 3 | 466 | 3832140 | ||
BC007352.2 | - | 3 | 514 | |||
BC010852.1 | - | 3 | 515 | 3898226 | ||
BC011586.1 | - | 3 | 466 | 4620484 | ||
BC012171.1 | - | 3 | 514 | 4642811 | ||
BC013996.2 | - | 3 | 470 | 3625337 | ||
BT007347.1 | - | 3 | 430 | |||
EU794591.1 | - | 3 | 539 | |||
Y16675.1 | - | 3 | 501 |