Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003693.3 | NP_003684.2 | 4, 5 | 889 | ||
NM_145350.2 | NP_663325.1 | 4, 5 | 889 | |||
NR_028075.2 | 4, 5 | 889 | ||||
NR_102409.1 | 4, 5 | 926 | ||||
GenBank mRNA | AB052947.1 | - | 4, 5 | 823 | ||
AB052948.1 | - | 4, 5 | 856 | |||
AB052949.1 | - | 4, 5 | 821 | |||
AB052950.1 | - | 4, 5 | 813 | |||
AK291810.1 | - | 4, 5 | 832 | |||
AK310174.1 | - | 4, 5 | 849 | |||
AK311370.1 | - | 2, 3 | 815 | |||
BC039735.1 | - | 4, 5 | 857 | 6041360 | ||
D63483.1 | - | 4, 5 | 799 | |||
D86864.1 | - | 4, 5 | 823 |