Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003718.4 | NP_003709.3 | 12 | 3388 | ||
NM_031267.3 | NP_112557.2 | 12 | 3388 | |||
XM_017012750.1 | XP_016868239.1 | 13 | 4337 | |||
XM_017012751.1 | XP_016868240.1 | 13 | 4337 | |||
GenBank mRNA | AB058694.1 | - | 6 | 2804 | ||
AJ297709.1 | - | 12 | 3238 | |||
AJ297710.1 | - | 12 | 3238 | |||
AK223053.1 | - | 6 | 541 | |||
BF707421.1 | - | 5 | 435 |