Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OASL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261825.1 | NP_001248754.1 | 2 | 557 | ||
NM_003733.3 | NP_003724.1 | 2 | 557 | |||
NM_198213.2 | NP_937856.1 | 2 | 557 | |||
XM_017020140.1 | XP_016875629.1 | 2 | 292 | |||
XM_017020141.1 | XP_016875630.1 | 2 | 292 | |||
GenBank mRNA | AJ225089.1 | - | 2 | 292 | ||
AK023956.1 | - | 2 | 293 | |||
AK314419.1 | - | 2 | 292 | |||
AL711263.1 | - | 2 | 295 | |||
BC117408.1 | - | 2 | 310 | 40125959 | ||
BC117410.1 | - | 2 | 310 | 40125961 | ||
BG339739.1 | - | 2 | 272 | |||
BG744291.1 | - | 2 | 240 | |||
JQ792168.1 | - | 2 | 286 |