Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBFA2T3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005187.5 | NP_005178.4 | 8 | 1431 | ||
NM_175931.2 | NP_787127.1 | 7 | 1097 | |||
XM_005256323.4 | XP_005256380.1 | 7 | 1409 | |||
XM_011523419.2 | XP_011521721.1 | 8 | 1533 | |||
XM_017023811.1 | XP_016879300.1 | 8 | 1533 | |||
GenBank mRNA | AB010419.1 | - | 8 | 1300 | ||
AB010420.1 | - | 7 | 1097 | |||
AF052220.1 | - | 2 | 123 | |||
AK308355.1 | - | 8 | 1030 | |||
AK314766.1 | - | 8 | 1286 | |||
BC047019.1 | - | 8 | 1281 | 5227349 | ||
BC062624.1 | - | 8 | 1301 | 5202447 |