Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083.3 | NP_001074.2 | 2 | 473 | ||
NM_033430.2 | NP_236914.2 | 2 | 304 | |||
NM_033437.3 | NP_246273.2 | 2 | 256 | |||
XM_017008791.1 | XP_016864280.1 | 2 | 727 | |||
GenBank mRNA | AB015656.1 | - | 2 | 198 | ||
AF043731.1 | - | 2 | 473 | |||
AF043732.1 | - | 2 | 660 | |||
AJ004865.1 | - | 2 | 411 | |||
AK290189.1 | - | 2 | 354 | |||
AY264918.1 | - | 2 | 192 | |||
BC126233.1 | - | 2 | 407 | 8991949 | ||
CD511199.1 | - | 2 | 221 | |||
JQ286346.1 | - | 2 | 318 |