Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083.3 | NP_001074.2 | 5, 6 | 1158 | ||
NM_033430.2 | NP_236914.2 | 5, 6 | 989 | |||
NM_033437.3 | NP_246273.2 | 5, 6 | 941 | |||
XM_017008791.1 | XP_016864280.1 | 5, 6 | 1412 | |||
GenBank mRNA | AB015656.1 | - | 5, 6 | 883 | ||
AF043731.1 | - | 5, 6 | 1158 | |||
AJ004865.1 | - | 5, 6 | 1096 | |||
AK290189.1 | - | 5, 6 | 1039 | |||
AY264918.1 | - | 5, 6 | 877 | |||
AY266363.1 | - | 4, 5 | 537 | |||
BC126233.1 | - | 5, 6 | 1092 | 8991949 | ||
JQ286346.1 | - | 5, 6 | 1003 |