Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284335.1 | NP_001271264.1 | 5 | 507 | ||
NM_001284336.1 | NP_001271265.1 | 3 | 360 | |||
NM_003758.3 | NP_003749.2 | 5 | 507 | |||
GenBank mRNA | AF090923.1 | - | 1 | 103 | ||
AK002006.1 | - | 5 | 427 | |||
AK023388.1 | - | 5 | 457 | |||
AK300659.1 | - | 5 | 392 | |||
AL536561.3 | - | 3 | 237 | |||
BC002719.2 | - | 5 | 440 | 3632801 | ||
U97670.1 | - | 5 | 440 |