Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003782.3 | NP_003773.1 | 1 | 1385 | ||
GenBank mRNA | AB026730.1 | - | 1 | 1356 | ||
AF063595.1 | - | 1 | 213 | |||
AK075312.1 | - | 1 | 1385 | |||
AK315647.1 | - | 1 | 1360 | |||
BC032574.1 | - | 1 | 1366 | 5247350 | ||
BU740293.1 | - | 1 | 316 | |||
CR457093.1 | - | 1 | 1105 | |||
Y15061.1 | - | 1 | 1105 |