Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003813.3 | NP_003804.2 | 2 | 2361 | ||
GenBank mRNA | AA758110.1 | - | 1 | 302 | ||
AF029900.1 | - | 1 | 1563 | |||
BC109024.1 | - | 1 | 2103 | 40027108 | ||
BC109025.1 | - | 1 | 2102 |