Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003823.3 | NP_003814.1 | 3 | 755 | ||
GenBank mRNA | AF104419.1 | - | 3 | 738 | ||
AF134240.1 | - | 3 | 638 | |||
AF217794.1 | - | 3 | 690 | |||
AY358279.1 | - | 3 | 738 | |||
BC017065.1 | - | 3 | 737 | 3886635 | ||
BC034349.1 | - | 3 | 746 | 4752507 |
Gene Symbol: RTEL1-TNFRSF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037882.1 | 38 | 5372 |