Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271572.1 | NP_001258501.1 | 3 | 545 | ||
NM_001271573.1 | NP_001258502.1 | 2 | 522 | |||
NM_003847.2 | NP_003838.1 | 3 | 638 | |||
GenBank mRNA | AB015594.1 | - | 3 | 581 | ||
AF093668.1 | - | 3 | 543 | |||
AK001415.1 | - | 3 | 631 | |||
AK297442.1 | - | 2 | 370 | |||
AK300978.1 | - | 3 | 423 | |||
AK313318.1 | - | 3 | 552 | |||
BC009697.1 | - | 3 | 532 | 3878406 | ||
BC101164.2 | - | 3 | 492 | 40015908 | ||
BC114384.1 | - | 3 | 492 | 40004153 | ||
CR542046.1 | - | 3 | 486 | |||
CR542075.1 | - | 3 | 486 |