Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003850.2 | NP_003841.1 | 7, 8 | 1010 | ||
XM_017020806.1 | XP_016876295.1 | 7, 8 | 1230 | |||
XM_017020807.1 | XP_016876296.1 | 9, 10 | 1245 | |||
XM_017020809.1 | XP_016876298.1 | 8, 9 | 1094 | |||
GenBank mRNA | AB035863.1 | - | 7, 8 | 1010 | ||
AF058953.1 | - | 6, 7 | 842 | |||
AF087890.1 | - | 7, 8 | 1018 | |||
AK001458.1 | - | 7, 8 | 908 | |||
AK001847.1 | - | 7, 8 | 997 | |||
AK225452.1 | - | 7, 8 | 997 | |||
AK299437.1 | - | 7, 8 | 949 | |||
AK302311.1 | - | 8, 9 | 1069 | |||
AK315513.1 | - | 7, 8 | 972 | |||
AK316041.1 | - | 8, 9 | 924 | |||
AK316271.1 | - | 8, 9 | 1325 | |||
AL080096.2 | - | 2, 3 | 199 | |||
BC027587.2 | - | 7, 8 | 962 | 4829291 | ||
BX640602.1 | - | 7, 8 | 981 | |||
KU178442.1 | - | 7, 8 | 953 | |||
KU178443.1 | - | 4, 5 | 509 |