Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317034.1 | NP_001303963.1 | 9 | 804 | ||
NM_001317035.1 | NP_001303964.1 | 9 | 780 | |||
NM_001317036.1 | NP_001303965.1 | 7 | 630 | |||
NM_003859.2 | NP_003850.1 | 8 | 699 | |||
NR_133648.1 | 8 | 721 | ||||
GenBank mRNA | AF007875.1 | - | 8 | 637 | ||
AK289569.1 | - | 8 | 667 | |||
BC007073.1 | - | 8 | 685 | 3997453 | ||
BC016322.1 | - | 8 | 670 | 4083041 | ||
BM922866.1 | - | 9 | 796 | |||
BQ441542.1 | - | 7 | 606 | |||
CA865473.1 | - | 2 | 410 | |||
CR456926.1 | - | 8 | 658 | |||
D86198.1 | - | 8 | 658 | |||
DR156925.1 | - | 9 | 759 | |||
KU178444.1 | - | 8 | 658 | |||
KU178445.1 | - | 9 | 739 | |||
KU178446.1 | - | 8 | 696 |