Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003862.2 | NP_003853.1 | 5 | 1091 | ||
GenBank mRNA | AB007422.1 | - | 5 | 554 | ||
AF075292.1 | - | 5 | 1091 | |||
AF211188.1 | - | 5 | 554 | |||
BC006245.2 | - | 5 | 698 | 3948893 | ||
BT019570.1 | - | 5 | 554 | |||
BT019571.1 | - | 5 | 554 |