Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006772.2 | NP_006763.2 | 9 | 1657 | ||
GenBank mRNA | AK094225.1 | - | 9 | 1657 | ||
AK307888.1 | - | 9 | 1472 | |||
AL713634.1 | - | 2 | 313 |