Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184879.1 | NP_001171808.1 | 2 | 451 | ||
NM_001184881.1 | NP_001171810.1 | 2 | 451 | |||
NM_003874.3 | NP_003865.1 | 2 | 451 | |||
XM_011510094.2 | XP_011508396.1 | 2 | 432 | |||
GenBank mRNA | AF054815.1 | - | 2 | 441 | ||
AF054816.1 | - | 2 | 441 | |||
AF054817.1 | - | 2 | 441 | |||
AF054818.1 | - | 2 | 441 | |||
AJ223324.1 | - | 2 | 416 | |||
AK313496.1 | - | 2 | 417 | |||
BC020063.1 | - | 2 | 396 | 4385965 | ||
CR541847.1 | - | 2 | 372 | |||
U82988.1 | - | 2 | 413 | |||
U96627.1 | - | 2 | 441 | |||
Y12632.1 | - | 2 | 441 |