Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014238.1 | NP_055053.1 | 21 | 2644 | ||
XM_006722154.3 | XP_006722217.1 | 18 | 2688 | |||
XM_011525429.2 | XP_011523731.1 | 20 | 2796 | |||
XM_011525430.2 | XP_011523732.1 | 20 | 2798 | |||
XM_011525431.2 | XP_011523733.1 | 20 | 2798 | |||
XM_017025267.1 | XP_016880756.1 | 19 | 2754 | |||
XM_017025268.1 | XP_016880757.1 | 19 | 2730 | |||
XM_017025269.1 | XP_016880758.1 | 19 | 2730 | |||
XM_017025270.1 | XP_016880759.1 | 19 | 2531 | |||
XM_017025271.1 | XP_016880760.1 | 21 | 3503 | |||
XM_017025272.1 | XP_016880761.1 | 21 | 3486 | |||
XM_017025273.1 | XP_016880762.1 | 20 | 3309 | |||
XM_017025274.1 | XP_016880763.1 | 20 | 2512 | |||
XM_017025275.1 | XP_016880764.1 | 20 | 2499 | |||
XM_017025276.1 | XP_016880765.1 | 21 | 3451 | |||
XM_017025277.1 | XP_016880766.1 | 16 | 2194 | |||
XM_017025278.1 | XP_016880767.1 | 16 | 2204 | |||
GenBank mRNA | U43586.1 | - | 16 | 1818 |