Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033121.1 | NP_149112.1 | 10 | 1217 | ||
XM_005253980.2 | XP_005254037.1 | 10 | 1260 | |||
XM_005253981.3 | XP_005254038.1 | 10 | 1217 | |||
XM_005253982.3 | XP_005254039.1 | 10 | 1214 | |||
XM_005253984.1 | XP_005254041.1 | 10 | 1016 | |||
XM_011538937.1 | XP_011537239.1 | 9 | 1116 | |||
XM_011538938.2 | XP_011537240.1 | 10 | 1220 | |||
XM_017020157.1 | XP_016875646.1 | 10 | 1220 | |||
XM_017020158.1 | XP_016875647.1 | 10 | 1214 | |||
XM_017020159.1 | XP_016875648.1 | 10 | 1000 | |||
XM_017020160.1 | XP_016875649.1 | 10 | 1000 | |||
XM_017020161.1 | XP_016875650.1 | 10 | 997 | |||
XM_017020162.1 | XP_016875651.1 | 10 | 1214 | |||
XM_017020163.1 | XP_016875652.1 | 9 | 1214 | |||
GenBank mRNA | AF064604.1 | - | 10 | 1123 | ||
AF155103.1 | - | 3 | 159 | |||
AK130522.1 | - | 1 | 626 | |||
AK293232.1 | - | 6 | 539 | |||
AK302535.1 | - | 10 | 1055 | |||
AK308923.1 | - | 10 | 1122 | |||
AY364251.1 | - | 10 | 958 | |||
BC032833.2 | - | 10 | 1058 | 4830174 |