Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCKBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318029.1 | NP_001304958.1 | 3 | 669 | ||
NM_176875.3 | NP_795344.1 | 4 | 921 | |||
XM_005253210.1 | XP_005253267.1 | 4 | 984 | |||
XM_017018516.1 | XP_016874005.1 | 4 | 1407 | |||
GenBank mRNA | AF239668.1 | - | 4 | 669 | ||
AF441129.1 | - | 4 | 666 | |||
AK226020.1 | - | 4 | 734 | |||
AK314266.1 | - | 4 | 699 | |||
AY029770.1 | - | 4 | 666 | |||
AY322551.1 | - | 4 | 666 | |||
BC000740.2 | - | 4 | 722 | 3504160 | ||
BT006789.1 | - | 4 | 666 | |||
CD013884.1 | - | 3 | 537 | |||
D13305.1 | - | 4 | 858 | |||
L04473.1 | - | 4 | 666 | |||
L07746.1 | - | 4 | 828 | |||
L08112.1 | - | 4 | 666 | |||
S70057.1 | - | 4 | 851 |
Set Membership: |
G-protein coupled receptor |