Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021095.2 | NP_066918.2 | 16 | 2167 | ||
NR_028323.1 | 15 | 2519 | ||||
XM_006712128.1 | XP_006712191.1 | 16 | 1962 | |||
XM_006712129.1 | XP_006712192.1 | 16 | 2010 | |||
XM_006712130.1 | XP_006712193.1 | 16 | 2001 | |||
XM_011533146.2 | XP_011531448.1 | 14 | 1727 | |||
XM_017005216.1 | XP_016860705.1 | 13 | 1737 | |||
GenBank mRNA | AF069307.1 | - | 16 | 2063 | ||
AF081571.1 | - | 15 | 1879 | |||
AK027855.1 | - | 15 | 2359 | |||
AK075078.1 | - | 7 | 605 | |||
AK293162.1 | - | 10 | 1051 | |||
AK314545.1 | - | 17 | 1994 | |||
AL096737.1 | - | 2 | 1221 | |||
BC012806.2 | - | 14 | 1729 | 3507776 | ||
BC015631.2 | - | 15 | 1880 | 3628126 | ||
BQ651664.1 | - | 4 | 406 |