Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAX1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079864.2 | NP_001073333.1 | 17 | 2346 | ||
NM_001206901.1 | NP_001193830.1 | 17 | 2349 | |||
NM_001206902.1 | NP_001193831.1 | 16 | 2158 | |||
NM_006024.6 | NP_006015.4 | 17 | 2472 | |||
XM_005249900.2 | XP_005249957.1 | 15 | 2151 | |||
XM_017012754.1 | XP_016868243.1 | 17 | 2442 | |||
GenBank mRNA | AF090891.1 | - | 17 | 2264 | ||
AF268075.1 | - | 16 | 2164 | |||
AK000098.1 | - | 17 | 2385 | |||
AK296720.1 | - | 16 | 2107 | |||
AK299729.1 | - | 17 | 2349 | |||
AK301064.1 | - | 17 | 2135 | |||
AK309757.1 | - | 6 | 560 | |||
AK314292.1 | - | 17 | 2442 | |||
AL136586.1 | - | 17 | 2378 | |||
BC050358.2 | - | 17 | 2357 | 6055189 | ||
CR457056.1 | - | 16 | 2164 | |||
CR533556.1 | - | 16 | 2290 | |||
U33821.2 | - | 17 | 2247 |