Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111045.1 | NP_001104515.1 | 4 | 965 | ||
NM_001111046.1 | NP_001104516.1 | 4 | 847 | |||
NM_001111047.1 | NP_001104517.1 | 4 | 844 | |||
NM_003914.3 | NP_003905.1 | 4 | 968 | |||
XM_011535294.2 | XP_011533596.1 | 4 | 1799 | |||
XM_011535295.2 | XP_011533597.1 | 4 | 1796 | |||
XM_011535296.2 | XP_011533598.1 | 4 | 624 | |||
GenBank mRNA | AK301897.1 | - | 4 | 740 | ||
AK314261.1 | - | 4 | 968 | |||
AK316392.1 | - | 4 | 767 | |||
BC035479.1 | - | 4 | 714 | |||
BC036346.1 | - | 4 | 714 | 5172478 | ||
BT019577.1 | - | 4 | 609 | |||
CD171793.1 | - | 4 | 767 | |||
U66838.1 | - | 4 | 747 | |||
U97680.1 | - | 3 | 457 |