Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003917.4 | NP_003908.1 | 2, 3 | 313 | ||
XM_005268167.3 | XP_005268224.1 | 2, 3 | 265 | |||
XM_005268168.4 | XP_005268225.1 | 2, 3 | 307 | |||
XM_005268169.3 | XP_005268226.1 | 2, 3 | 264 | |||
XM_005268170.3 | XP_005268227.1 | 2, 3 | 264 | |||
XM_005268172.3 | XP_005268229.1 | 2, 3 | 264 | |||
XM_005268173.3 | XP_005268230.1 | 2, 3 | 264 | |||
XM_005268180.4 | XP_005268237.1 | 1, 2 | 489 | |||
XM_005268182.3 | XP_005268239.1 | 2, 3 | 235 | |||
XM_006720301.3 | XP_006720364.1 | 2, 3 | 362 | |||
XM_011537283.2 | XP_011535585.1 | 1, 2 | 211 | |||
XM_017021740.1 | XP_016877229.1 | 1, 2 | 211 | |||
XM_017021743.1 | XP_016877232.1 | 1, 2 | 211 | |||
XM_017021744.1 | XP_016877233.1 | 2, 3 | 265 | |||
XM_017021745.1 | XP_016877234.1 | 2, 3 | 265 | |||
XM_017021746.1 | XP_016877235.1 | 2, 3 | 235 | |||
GenBank mRNA | AB015318.1 | - | 2, 3 | 255 | ||
AF068706.1 | - | 1, 2 | 973 | |||
AK309103.1 | - | 2, 3 | 265 | |||
BG701521.1 | - | 2, 3 | 316 | |||
BI918958.1 | - | 2, 3 | 319 |