Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003406.1 | NP_001003406.1 | 31 | 4995 | ||
NM_021096.3 | NP_066919.2 | 32 | 5100 | |||
XM_017029035.1 | XP_016884524.1 | 22 | 8370 | |||
XM_017029036.1 | XP_016884525.1 | 22 | 8370 | |||
XM_017029037.1 | XP_016884526.1 | 22 | 8371 | |||
GenBank mRNA | AB032946.1 | - | 13 | 1504 | ||
AF129133.1 | - | 32 | 5100 | |||
AF142567.1 | - | 32 | 5100 | |||
AF211189.1 | - | 31 | 4995 | |||
AF393329.1 | - | 31 | 4995 |