Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282112.1 | NP_001269041.1 | 6, 7 | 794 | ||
NM_001282113.1 | NP_001269042.1 | 6, 7 | 787 | |||
NM_003935.4 | NP_003926.1 | 6, 7 | 1110 | |||
XM_005261811.1 | XP_005261868.1 | 8, 9 | 960 | |||
XM_005261813.2 | XP_005261870.1 | 7, 8 | 825 | |||
XM_006724349.1 | XP_006724412.1 | 7, 8 | 771 | |||
XM_006724350.1 | XP_006724413.1 | 7, 8 | 945 | |||
XM_011530482.1 | XP_011528784.1 | 7, 8 | 1027 | |||
XM_017029038.1 | XP_016884527.1 | 6, 7 | 773 | |||
XM_017029039.1 | XP_016884528.1 | 8, 9 | 835 | |||
XM_017029040.1 | XP_016884529.1 | 7, 8 | 811 | |||
XM_017029041.1 | XP_016884530.1 | 8, 9 | 853 | |||
GenBank mRNA | AF017146.1 | - | 5, 6 | 695 | ||
AF053082.1 | - | 5, 6 | 695 | |||
AF125216.1 | - | 5, 6 | 655 | |||
AK290997.1 | - | 6, 7 | 740 | |||
AL833505.1 | - | 7, 8 | 824 | |||
BC002432.2 | - | 6, 7 | 1017 | 3346377 | ||
CR456596.1 | - | 5, 6 | 609 |