Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136017.3 | NP_001129489.1 | 3 | 661 | ||
NM_001136125.2 | NP_001129597.1 | 2 | 448 | |||
NM_001136126.2 | NP_001129598.1 | 2 | 445 | |||
NM_001287427.1 | NP_001274356.1 | 3 | 1164 | |||
NM_001287434.1 | NP_001274363.1 | 3 | 627 | |||
NM_001760.4 | NP_001751.1 | 3 | 664 | |||
XM_011514971.1 | XP_011513273.1 | 3 | 659 | |||
XM_011514972.1 | XP_011513274.1 | 3 | 455 | |||
GenBank mRNA | AB209825.1 | - | 3 | 455 | ||
AK055519.1 | - | 3 | 722 | |||
AK057206.1 | - | 3 | 455 | |||
AK096276.1 | - | 3 | 975 | |||
AK097856.1 | - | 2 | 239 | |||
AK299187.1 | - | 2 | 366 | |||
AK303452.1 | - | 3 | 443 | |||
AK315421.1 | - | 3 | 580 | |||
BC011616.2 | - | 3 | 546 | 3946312 | ||
CR542246.1 | - | 3 | 465 | |||
DA069566.1 | - | 2 | 264 | |||
DA999482.1 | - | 2 | 445 | |||
DB001987.1 | - | 2 | 423 | |||
DB110132.1 | - | 3 | 421 | |||
DC360231.1 | - | 2 | 355 | |||
M90814.1 | - | 3 | 565 | |||
M92287.1 | - | 3 | 630 |