Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171156.1 | NP_001164627.1 | 11 | 1996 | ||
NM_001171157.1 | NP_001164628.1 | 10 | 1885 | |||
NM_001171158.1 | NP_001164629.1 | 10 | 1741 | |||
NM_001171159.1 | NP_001164630.1 | 10 | 1711 | |||
NM_001171161.1 | NP_001164632.1 | 9 | 1441 | |||
NM_001322105.1 | NP_001309034.1 | 9 | 1615 | |||
NM_033130.4 | NP_149121.2 | 11 | 2170 | |||
XM_005259366.4 | XP_005259423.1 | 10 | 1728 | |||
XM_005259367.3 | XP_005259424.1 | 10 | 1559 | |||
XM_011527439.1 | XP_011525741.1 | 11 | 1854 | |||
GenBank mRNA | AF301007.1 | - | 11 | 2065 | ||
AF310233.1 | - | 11 | 1985 | |||
AF311905.1 | - | 10 | 1885 | |||
AK122619.1 | - | 8 | 1151 | |||
AK289900.1 | - | 10 | 1671 | |||
AK290542.1 | - | 10 | 1544 | |||
AK292025.1 | - | 10 | 1788 | |||
AK294185.1 | - | 10 | 1475 | |||
AK303514.1 | - | 10 | 1580 | |||
AK303717.1 | - | 9 | 1454 | |||
AK308982.1 | - | 8 | 1551 | |||
AY032685.1 | - | 11 | 1930 | |||
AY358337.1 | - | 10 | 1534 | |||
BC009955.2 | - | 7 | 940 | |||
BC101725.1 | - | 11 | 2035 | 8069231 | ||
BC143497.1 | - | 10 | 1750 | 9052005 | ||
BC143500.1 | - | 11 | 1861 | 9052008 |