Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MVB12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011703.2 | NP_001011703.1 | 4 | 461 | ||
NM_033446.2 | NP_258257.1 | 4 | 461 | |||
XM_005252297.1 | XP_005252354.1 | 4 | 496 | |||
XM_017015275.1 | XP_016870764.1 | 4 | 461 | |||
XM_017015276.1 | XP_016870765.1 | 4 | 496 | |||
XM_017015277.1 | XP_016870766.1 | 4 | 461 | |||
XM_017015278.1 | XP_016870767.1 | 4 | 461 | |||
XM_017015279.1 | XP_016870768.1 | 3 | 316 | |||
GenBank mRNA | AK024432.1 | - | 4 | 456 | ||
AK293740.1 | - | 4 | 455 | |||
AK298023.1 | - | 3 | 357 | |||
AK307473.1 | - | 3 | 293 | |||
BC000122.1 | - | 4 | 436 | |||
BC028675.1 | - | 4 | 447 | 4083276 | ||
DA339911.1 | - | 4 | 461 |