Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006944.1 | NP_001006945.1 | 2 | 179 | ||
NM_001300802.1 | NP_001287731.1 | 2 | 179 | |||
NM_003942.2 | NP_003933.1 | 2 | 179 | |||
XM_005274380.1 | XP_005274437.1 | 2 | 159 | |||
GenBank mRNA | AF074715.1 | - | 1 | 48 | ||
AJ010119.1 | - | 2 | 161 | |||
AK223561.1 | - | 2 | 184 | |||
AK292163.1 | - | 2 | 160 | |||
AK313567.1 | - | 2 | 159 | |||
BC028079.1 | - | 2 | 143 | |||
BC047896.1 | - | 2 | 158 | 5763859 | ||
BI253655.1 | - | 2 | 179 | |||
BI523588.1 | - | 2 | 204 |