Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006944.1 | NP_001006945.1 | 3 | 410 | ||
NM_001300802.1 | NP_001287731.1 | 3 | 410 | |||
NM_001318361.1 | NP_001305290.1 | 3 | 364 | |||
NM_003942.2 | NP_003933.1 | 3 | 410 | |||
XM_005274380.1 | XP_005274437.1 | 3 | 390 | |||
GenBank mRNA | AF074715.1 | - | 2 | 279 | ||
AJ010119.1 | - | 3 | 392 | |||
AK223561.1 | - | 3 | 415 | |||
AK292163.1 | - | 3 | 391 | |||
AK298507.1 | - | 3 | 335 | |||
AK313567.1 | - | 3 | 390 | |||
BC028079.1 | - | 3 | 374 | |||
BC047896.1 | - | 3 | 389 | 5763859 | ||
BI253655.1 | - | 3 | 410 | |||
BI523588.1 | - | 3 | 435 |