Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171170.1 | NP_001164641.1 | 6 | 685 | ||
NM_030631.3 | NP_085134.1 | 6 | 685 | |||
XM_011537287.2 | XP_011535589.1 | 7 | 467 | |||
XM_011537288.2 | XP_011535590.1 | 6 | 559 | |||
XM_011537289.2 | XP_011535591.1 | 5 | 468 | |||
GenBank mRNA | AJ278148.1 | - | 6 | 685 | ||
AK289575.1 | - | 6 | 448 | |||
BC101521.1 | - | 6 | 421 | 8069027 | ||
BC113365.1 | - | 6 | 421 | 8322417 |