Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033343.3 | NP_203129.1 | 5, 6 | 1016 | ||
XM_011510105.2 | XP_011508407.1 | 5, 6 | 1250 | |||
XM_011510106.2 | XP_011508408.1 | 5, 6 | 873 | |||
XM_011510108.2 | XP_011508410.1 | 5, 6 | 656 | |||
XM_017002755.1 | XP_016858244.1 | 4, 5 | 1368 | |||
GenBank mRNA | AB037683.1 | - | 2, 3 | 334 | ||
AB055703.1 | - | 4, 5 | 719 | |||
AF179849.1 | - | 5, 6 | 1020 | |||
AF282899.1 | - | 4, 5 | 715 | |||
AY053457.1 | - | 5, 6 | 808 | |||
BC011759.2 | - | 5, 6 | 827 | 3529089 | ||
KU178818.1 | - | 5, 6 | 784 | |||
KU178819.1 | - | 6, 7 | 835 |