Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052844.3 | NP_443076.2 | 2 | 446 | ||
XM_011519179.2 | XP_011517481.1 | 2 | 380 | |||
GenBank mRNA | AK302098.1 | - | 1 | 76 | ||
AL555205.3 | - | 2 | 253 | |||
BC001614.1 | - | 2 | 281 | |||
BG177634.1 | - | 2 | 289 |