Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099437.1 | NP_001092907.1 | 5 | 2034 | ||
NM_001099438.1 | NP_001092908.1 | 5 | 2100 | |||
NM_194325.2 | NP_919306.2 | 5 | 2031 | |||
NR_024018.1 | 6 | 2111 | ||||
XM_011527443.2 | XP_011525745.1 | 5 | 2150 | |||
XM_011527444.1 | XP_011525746.1 | 5 | 1739 | |||
XM_017027423.1 | XP_016882912.1 | 4 | 2359 | |||
XM_017027424.1 | XP_016882913.1 | 5 | 1785 | |||
XM_017027425.1 | XP_016882914.1 | 5 | 2090 | |||
XM_017027426.1 | XP_016882915.1 | 5 | 2546 | |||
XM_017027427.1 | XP_016882916.1 | 6 | 2126 | |||
XM_017027428.1 | XP_016882917.1 | 6 | 2130 | |||
XM_017027429.1 | XP_016882918.1 | 6 | 2157 | |||
GenBank mRNA | AK000569.1 | - | 1 | 1184 | ||
AK290435.1 | - | 6 | 1966 | |||
AK295515.1 | - | 5 | 1890 | |||
BC142996.1 | - | 5 | 2019 | 8860451 | ||
BX640666.1 | - | 6 | 2034 |