Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256526.1 | NP_001243455.1 | 2 | 601 | ||
NM_152833.2 | NP_690046.3 | 2 | 642 | |||
NR_134506.1 | 2 | 640 | ||||
GenBank mRNA | AK023162.1 | - | 2 | 593 | ||
AK091349.1 | - | 2 | 373 | |||
BC013282.2 | - | 2 | 581 | |||
BC014304.2 | - | 2 | 539 | |||
BC021231.2 | - | 2 | 620 | |||
BC064970.1 | - | 1 | 155 | |||
BC084549.1 | - | 1 | 222 | |||
BC092490.1 | - | 2 | 291 | |||
BC111053.1 | - | 2 | 347 |