Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIP1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303097.1 | NP_001290026.1 | 6 | 579 | ||
NM_001303099.1 | NP_001290028.1 | 6 | 665 | |||
NM_003959.2 | NP_003950.1 | 6 | 579 | |||
XM_011538963.1 | XP_011537265.1 | 6 | 882 | |||
GenBank mRNA | AB014555.1 | - | 6 | 513 | ||
AK090537.1 | - | 6 | 632 | |||
AK298387.1 | - | 6 | 536 | |||
BC067085.1 | - | 6 | 528 | 6085487 |