Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53I13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138349.2 | NP_612358.3 | 6 | 1031 | ||
XM_017025282.1 | XP_016880771.1 | 6 | 1403 | |||
GenBank mRNA | AB110209.1 | - | 6 | 1098 | ||
AK075341.1 | - | 6 | 1062 | |||
AK093030.1 | - | 1 | 1933 | |||
AL137404.1 | - | 1 | 301 | |||
BC001593.2 | - | 6 | 1031 | 2988166 | ||
BC004507.2 | - | 1 | 403 |