Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER1L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293083.1 | NP_001280012.1 | 37, 38 | 4188 | ||
XM_011512110.2 | XP_011510412.1 | 37, 38 | 4205 | |||
XM_011512111.2 | XP_011510413.1 | 37, 38 | 4202 | |||
XM_011512113.2 | XP_011510415.1 | 37, 38 | 4199 | |||
XM_011512114.2 | XP_011510416.1 | 37, 38 | 4202 | |||
XM_011512115.2 | XP_011510417.1 | 36, 37 | 4136 | |||
XM_011512116.2 | XP_011510418.1 | 37, 38 | 4118 | |||
XM_011512117.2 | XP_011510419.1 | 36, 37 | 4100 | |||
XM_011512118.2 | XP_011510420.1 | 36, 37 | 4085 | |||
XM_011512119.2 | XP_011510421.1 | 37, 38 | 4205 | |||
XM_011512120.2 | XP_011510422.1 | 36, 37 | 4096 | |||
XM_011512122.2 | XP_011510424.1 | 37, 38 | 4206 | |||
XM_011512124.2 | XP_011510426.1 | 21, 22 | 3789 | |||
XM_011512125.2 | XP_011510427.1 | 37, 38 | 4206 | |||
XM_017005222.1 | XP_016860711.1 | 37, 38 | 4202 | |||
XM_017005223.1 | XP_016860712.1 | 20, 21 | 2853 | |||
XM_017005224.1 | XP_016860713.1 | 11, 12 | 1645 | |||
GenBank mRNA | AL133022.1 | - | 19, 20 | 3591 | ||
AY461813.1 | - | 36, 37 | 4217 | |||
BC117324.1 | - | 3, 4 | 396 | |||
BC126368.1 | - | 3, 4 | 399 | |||
BC143825.1 | - | 3, 4 | 396 |