Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317800.1 | NP_001304729.1 | 3 | 440 | ||
NM_003974.3 | NP_003965.2 | 5 | 810 | |||
NM_201349.2 | NP_958728.1 | 4 | 722 | |||
XM_005273680.4 | XP_005273737.1 | 4 | 537 | |||
GenBank mRNA | AF034970.1 | - | 5 | 703 | ||
AK226110.1 | - | 3 | 410 | |||
AK292126.1 | - | 5 | 766 | |||
AK296671.1 | - | 4 | 704 | |||
AL545478.3 | - | 1 | 962 | |||
BC032623.1 | - | 5 | 778 | 5242017 | ||
CB270269.1 | - | 3 | 394 |