Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSTPIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321135.1 | NP_001308064.1 | 12 | 1399 | ||
NM_001321136.1 | NP_001308065.1 | 14 | 1675 | |||
NM_001321137.1 | NP_001308066.1 | 14 | 1272 | |||
NM_003978.4 | NP_003969.2 | 13 | 1456 | |||
NR_135552.1 | 10 | 1188 | ||||
XM_006720737.2 | XP_006720800.1 | 13 | 1188 | |||
XM_011522163.1 | XP_011520465.1 | 14 | 1474 | |||
XM_011522164.1 | XP_011520466.1 | 14 | 1142 | |||
XM_011522165.2 | XP_011520467.1 | 13 | 997 | |||
XM_011522166.1 | XP_011520468.1 | 14 | 1508 | |||
XM_011522167.1 | XP_011520469.1 | 13 | 1383 | |||
XM_011522168.2 | XP_011520470.1 | 14 | 1474 | |||
XM_011522169.1 | XP_011520471.1 | 12 | 1286 | |||
GenBank mRNA | AB451310.1 | - | 13 | 967 | ||
AB451440.1 | - | 13 | 967 | |||
AF038602.1 | - | 12 | 910 | |||
AF038603.1 | - | 13 | 967 | |||
AK308462.1 | - | 14 | 1306 | |||
AK309856.1 | - | 14 | 1638 | |||
AL110110.1 | - | 2 | 116 | |||
BC008602.1 | - | 13 | 1328 | 4180398 | ||
KU178458.1 | - | 13 | 967 | |||
KU178459.1 | - | 10 | 699 | |||
U94778.1 | - | 13 | 1183 |