Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005443.4 | NP_005434.4 | 3 | 458 | ||
XM_011532400.1 | XP_011530702.1 | 3 | 438 | |||
XM_011532401.1 | XP_011530703.1 | 3 | 524 | |||
GenBank mRNA | AF016496.1 | - | 3 | 374 | ||
AF033026.1 | - | 3 | 400 | |||
AF105227.1 | - | 3 | 428 | |||
AK292774.1 | - | 3 | 408 | |||
AK297981.1 | - | 3 | 408 | |||
AU132232.1 | - | 3 | 458 | |||
BC011392.1 | - | 3 | 331 | 3869484 | ||
BC032513.1 | - | 3 | 427 | |||
BC040940.1 | - | 3 | 386 | |||
BC050627.1 | - | 3 | 394 | 5556165 | ||
BQ437924.1 | - | 3 | 386 | |||
CR457028.1 | - | 3 | 374 | |||
Y10387.1 | - | 3 | 410 |