Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005443.4 | NP_005434.4 | 4 | 523 | ||
XM_011532400.1 | XP_011530702.1 | 4 | 503 | |||
XM_011532401.1 | XP_011530703.1 | 4 | 589 | |||
GenBank mRNA | AF016496.1 | - | 4 | 439 | ||
AF033026.1 | - | 4 | 465 | |||
AF105227.1 | - | 4 | 493 | |||
AK292774.1 | - | 4 | 473 | |||
AU132232.1 | - | 4 | 523 | |||
BC011392.1 | - | 4 | 396 | 3869484 | ||
BC032513.1 | - | 4 | 492 | |||
BC040940.1 | - | 4 | 451 | |||
BC050627.1 | - | 4 | 459 | 5556165 | ||
BQ437924.1 | - | 4 | 451 | |||
CR457028.1 | - | 4 | 439 | |||
Y10387.1 | - | 4 | 475 |