Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4BP2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079691.1 | NP_001073159.1 | 1 | 216 | ||
NM_001286459.1 | NP_001273388.1 | 1 | 216 | |||
NM_001286460.1 | NP_001273389.1 | 1 | 216 | |||
NM_001286461.1 | NP_001273390.1 | 1 | 216 | |||
NM_052818.2 | NP_438169.2 | 1 | 216 | |||
XM_006719890.3 | XP_006719953.1 | 1 | 472 | |||
XM_011535302.2 | XP_011533604.1 | 1 | 168 | |||
XM_017020836.1 | XP_016876325.1 | 1 | 1671 | |||
XM_017020837.1 | XP_016876326.1 | 1 | 456 | |||
XM_017020839.1 | XP_016876328.1 | 1 | 1687 | |||
XM_017020840.1 | XP_016876329.1 | 1 | 195 | |||
XM_017020842.1 | XP_016876331.1 | 1 | 168 | |||
GenBank mRNA | AL049785.1 | - | 1 | 172 | ||
AL049786.1 | - | 1 | 172 | |||
AL049787.1 | - | 1 | 172 | |||
BC022188.2 | - | 1 | 162 | |||
BF448179.1 | - | 1 | 53 | |||
BI551105.1 | - | 1 | 221 | |||
U50527.1 | - | 1 | 167 |