Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021195.4 | NP_067018.2 | 2 | 650 | ||
GenBank mRNA | AF125306.1 | - | 1 | 590 | ||
AK075329.1 | - | 2 | 648 | |||
AY358480.1 | - | 1 | 595 | |||
BC008934.2 | - | 1 | 624 | 2963008 | ||
BT007399.1 | - | 1 | 590 | |||
BX444798.2 | - | 2 | 649 |