Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021101.4 | NP_066924.1 | 2 | 588 | ||
GenBank mRNA | AA484795.1 | - | 3 | 339 | ||
AF101051.1 | - | 2 | 568 | |||
AF114837.1 | - | 2 | 577 | |||
AF115546.1 | - | 2 | 358 | |||
AF134160.1 | - | 2 | 595 | |||
AK296934.1 | - | 3 | 469 | |||
AK312866.1 | - | 2 | 584 | |||
AY358652.1 | - | 2 | 566 | |||
BC012471.1 | - | 2 | 546 | 4500534 | ||
CR457138.1 | - | 2 | 348 | |||
EF564137.1 | - | 2 | 348 |