Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316990.1 | NP_001303919.1 | 7 | 988 | ||
NM_138363.2 | NP_612372.1 | 8 | 1094 | |||
NR_133644.1 | 8 | 1113 | ||||
XM_005257779.3 | XP_005257836.1 | 8 | 1070 | |||
XM_005257780.2 | XP_005257837.1 | 7 | 713 | |||
XM_006722166.3 | XP_006722229.1 | 8 | 852 | |||
XM_011525458.2 | XP_011523760.1 | 8 | 1070 | |||
XM_017025287.1 | XP_016880776.1 | 8 | 2073 | |||
XM_017025288.1 | XP_016880777.1 | 7 | 746 | |||
GenBank mRNA | AK057326.1 | - | 8 | 1068 | ||
AK290587.1 | - | 8 | 834 | |||
AK297409.1 | - | 7 | 689 | |||
AK316257.1 | - | 7 | 732 | |||
AL832822.1 | - | 8 | 764 | |||
BC009518.1 | - | 8 | 790 | 3633884 |