Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM55B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100814.2 | NP_001094284.1 | 5, 6 | 903 | ||
NM_144568.3 | NP_653169.2 | 5, 6 | 882 | |||
GenBank mRNA | AK314021.1 | - | 5, 6 | 883 | ||
AL137727.1 | - | 1, 2 | 2010 | |||
BC002867.1 | - | 5, 6 | 664 | 3940519 | ||
BC020947.1 | - | 5, 6 | 747 | 4816227 | ||
BX161490.1 | - | 5, 6 | 748 | |||
BX248025.1 | - | 5, 6 | 712 |